Anti-MAP3K20 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A39364-50UG
77806-154EA 225 USD
77806-154
Anti-MAP3K20 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500 to 1:2000, ELISA:1:40000

Type:
Antigen: MAP3K20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR