Anti-NDUFS5 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A39234-100UG A39234-50UG
77805-953EA 330 USD
77805-953 77805-954
Anti-NDUFS5 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: NDUFS5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR