Anti-ZNF596 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A39069-100UG A39069-50UG
77805-655EA 330 USD
77805-655 77805-656
Anti-ZNF596 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: ZNF596
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR