Anti-ZNF529 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A39009-50UG
77805-542EA 225 USD
77805-542
Anti-ZNF529 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200

Type:
Antigen: ZNF529
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR