Anti-ZNF513 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A39007-100UG A39007-50UG
77805-538EA 330 USD
77805-538 77805-539
Anti-ZNF513 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: ZNF513
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR