Anti-ZNF232 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A38819-100UG A38819-50UG
77805-191EA 330 USD
77805-191 77805-192
Anti-ZNF232 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: ZNF232
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR