Anti-ZBTB40 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A38645-100UL
77804-887EA 585 USD
77804-887
Anti-ZBTB40 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: ZBTB40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR