Anti-VDAC2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A38278-100UG A38278-50UG
77804-321EA 330 USD
77804-321 77804-322
Anti-VDAC2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: VDAC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR