Anti-UGGT1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A38010-100UL
77803-953EA 585 USD
77803-953
Anti-UGGT1 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: UGGT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR