Anti-TPM3 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A37302-100UG A37302-50UG
77811-379EA 330 USD
77811-379 77803-379
Anti-TPM3 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TPM3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR