Anti-TIPIN Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A36631-100UL
77802-618EA 585 USD
77802-618
Anti-TIPIN Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TIPIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR