Anti-THRA Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A36562-50UG A36562-100UG
77802-514EA 225 USD
77802-514 77802-513
Anti-THRA Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: THRA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR