Anti-THRA Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A36559-100UL
77802-508EA 585 USD
77802-508
Anti-THRA Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: THRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR