Anti-Thbs3 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A36542-100UG A36542-50UG
77802-484EA 330 USD
77802-484 77802-485
Anti-Thbs3 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: Thbs3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR