Anti-TGFB3 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A36490-100UG A36490-50UG
77802-398EA 330 USD
77802-398 77802-399
Anti-TGFB3 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: TGFB3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR