Anti-SULT2A1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A35981-100UL A35981-50UL
77801-572EA 330 USD
77801-572 77801-573
Anti-SULT2A1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: SULT2A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR