Anti-SSX5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A35716-100UL
77801-158EA 585 USD
77801-158
Anti-SSX5 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: SSX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR