Anti-SOX10 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A35424-100UL
77800-695EA 585 USD
77800-695
Anti-SOX10 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: SOX10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR