Anti-SNX24 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A35338-100UG A35338-50UG
77800-552EA 330 USD
77800-552 77800-553
Anti-SNX24 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: SNX24
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR