Anti-SERPINA9 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A35288-100UL A35288-50UL
77800-473EA 330 USD
77800-473 77800-474
Anti-SERPINA9 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100

Type:
Antigen: SERPINA9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR