Anti-SMYD4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A35239-100UL
77800-394EA 585 USD
77800-394
Anti-SMYD4 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: SMYD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR