Anti-SLC34A2 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A35001-100UG A35001-50UG
77800-015EA 330 USD
77800-015 77800-016
Anti-SLC34A2 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: SLC34A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR