Anti-SLC25A5 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A34947-50UG A34947-100UG
77799-924EA 225 USD
77799-924 77799-923
Anti-SLC25A5 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SLC25A5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR