Anti-SLC25A14 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A34926-100UG A34926-50UG
77799-894EA 330 USD
77799-894 77799-895
Anti-SLC25A14 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SLC25A14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR