Anti-SIN3B Rabbit Recombinant Antibody

Supplier: AFG Bioscience
A34734-100UG A34734-50UG
77799-587EA 330 USD
77799-587 77799-588
Anti-SIN3B Rabbit Recombinant Antibody
Antibodies
Recommended dilution: IHC: 1:200 to 1:500, IF: 1:50 to 1:200

Type:
Antigen: SIN3B
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR