Anti-SAA1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A34108-100UG A34108-50UG
77798-602EA 330 USD
77798-602 77798-603
Anti-SAA1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: SAA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR