Anti-RGS22 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A33167-100UG A33167-50UG
77797-217EA 330 USD
77797-217 77797-218
Anti-RGS22 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: RGS22
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR