Anti-PYGL Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A32552-100UG A32552-50UG
77796-262EA 330 USD
77796-262 77796-263
Anti-PYGL Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: PYGL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR