Anti-PTPRZ1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A32491-100UG A32491-50UG
77796-165EA 330 USD
77796-165 77796-166
Anti-PTPRZ1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20 to 1:200

Type:
Antigen: PTPRZ1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR