Anti-PRKDC Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A32019-100UG A32019-50UG
77795-396EA 330 USD
77795-396 77795-397
Anti-PRKDC Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: PRKDC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR