Anti-PRIM1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A31975-100UG A31975-50UG
77795-332EA 330 USD
77795-332 77795-333
Anti-PRIM1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: PRIM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR