Anti-POMP Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A31646-100UG A31646-50UG
77794-820EA 330 USD
77794-820 77794-821
Anti-POMP Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: POMP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR