Anti-POLR1C Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A31589-100UG A31589-50UG
77794-726EA 330 USD
77794-726 77794-727
Anti-POLR1C Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: POLR1C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR