Platinum® Pro Instrument, Quantum-Si
Supplier: QUANTUM-SI
910-10033-00
77778-306EA 120000 USD
77778-306
Platinum® Pro Instrument, Quantum-Si
Next Generation Sequencing Instruments
Platinum Pro Instrument for next-generation protein sequencing™.
  • Accessible benchtop instrument enables protein sequencing with limited laboratory space
  • Intuitive touch screen interface unlocks advanced protein analysis for novice and experienced users
  • Scalable solution delivers sample insights with less than three hours hands on time and regular innovative enhancements
  • Versatile with cloud analysis and storage or local informatics option
  • Insight generating solution for divers protein applications with single-molecule amino acid resolution including: Protein identification, Protein variant and post translational modification analysis, antibody sequencing, and protein screening with barcoding
Platinum Pro makes next-generation protein sequencing accessible today and for the future, packed with capability across diverse applications. Platinum Pro opens access to single molecule insights for every lab, everywhere.
Certifications:
Conforms to ICES-003:2020 Issue 7, UL STD(s) 61010-1 & 61010-2-081, and is certified to CSA STD C22.2# 61010-1-12 & 61010-2-081. 
Conforms with low voltage directives 2014/35/EC; EMC Directive 2014/35/EU; ROHS Directive 2011/65/EU; WEEE Directive 2012/19/EU.    
Conformité Européenne (CE) Assessment meets requirements for all relevant EU and UK directives. Meets Waste Electrical and Electronic Equipment Directive (WEEE).
Ordering information:
Includes 12 months manufacturer's warranty including Platinum Analysis Software and cloud data storage.