PYREX® Parr Weighing Bottle, Corning
Supplier: Corning
1684-2024
77776-950CS 671.25 USD
77776-950
PYREX® Parr Weighing Bottle, Corning
Bottles Weighing Bottles
PYREX® borosilicate glass, cylindrical.
  • Excellent chemical resistance
  • Excellent temperature resistance
These Parr weighing bottles are designed for weighing small samples and have an external 24/12 standard taper joint style opening. The closure style cap fits over the external ground body.