SP Bel-Art Electrophoresis Gel Plate Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F13596-0000
77776-794EA 185.84 USD
77776-794
SP Bel-Art Electrophoresis Gel Plate Rack, Bel-Art Products, a part of SP
Racks Electrophoresis Plate Racks
This “A” shaped rack has ten places for holding glass plates and a weighted base for safety.
  • Polypropylene construction cushions glass edges
  • Secure transport without chipping or cracking
  • Small footprint
  • Weighted base to prevent tipping over
It features a round bottom and wide handles to provide a secure grip while transporting the rack. Smooth polypropylene material cushions glass edges to reduce chipping and cracking. A flat area on each end is designed for labels. The rack has a footprint of 20.3Wx38.1Dx22.7H cm (8x15x9") and weighs 1.7kg (33/4lbs). Available in white with steel fittings.