VWR® Traceable® Snap-In Module Thermometer with Probe
Supplier: VWR International
77776-730 10171-280 10171-444 10171-560
77776-730EA 63.93 USD
77776-730 10171-280 10171-444 10171-560
VWR® Traceable® Snap-In Module Thermometer with Probe
Thermometers Digital Thermometers
This versatile panel-mount module snaps into place or is a stand-alone unit.
  • Traceable®
  • Reads in °C and °F
  • Memory function stores minimum/maximum readings
It comes complete with a stainless steel probe featuring an extended 3m (10') cable with handle. The module records minimum and maximum memory readings over any time period. The easy-view, four-digit LCD measures 1.9 cm (3/4") high.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.