VWR® Traceable® Digital Thermometer with Recorder Output
Supplier: VWR International
90028-89
77776-726EA 283.83 USD
77776-726
VWR® Traceable® Digital Thermometer with Recorder Output
Thermometers Digital Thermometers
This thermometer’s recorder output jack provides a 1mV signal per degree, which enables it to be used with any chart recorder, strip recorder, or computer that accepts millivolt signals.
  • Traceable®
  • Stores highest readings
  • Reads °C and °F
  • Provides a 1 millivolt signal per degree
Ideal for monitoring conditions in chambers, cleanrooms, food processing, scientific labs, and all temperature-sensitive manufacturing operations.
The maximum button captures the highest reading achieved. The hold button freezes the display to capture a current reading. Uses all Type-K thermocouples. Unit has a large, bright, 1.3 cm (1/2"), four-digit LCD display.
All Type-K probes may be used with liquids, air/gas, and semi-solids. Stainless steel probe with handle has a 15.9 cm (61/4") stem with a 3.3mm (1/8") diameter fitted to an 8.9 cm (31/2") handle. Beaded probe is 1.5mm (1/16") in diameter and features a 1.2m (4') fluoropolymer resin cable. The low-temperature stainless steel probe has a diameter of 4.3mm (3/16") and features a 30.5 cm (12") stem with a 12.7 cm (5") handle. Low-temperature probe is supplied with a 2.4m (8') expandable cable. High-temperature probe features 3m (10') braided metal wire cable with a smooth tip and measures 5mm (3/16") in diameter.
Signal output: 1 mV per degree in 0.1°C display mode, 0.1 mV per degree in 1°C display mode
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate thermometer traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a fast-response Type-K beaded probe (61161-372), a 9V battery, and a recorder output jack.