VWR® Memory Wide-Range Thermometer
Supplier: VWR International
77776-716
77776-716EA 957.72 USD
77776-716
VWR® Memory Wide-Range Thermometer
Thermometers Digital Thermometers
Unit features red backlighting of the display so that answers may be read in poor lighting or darkness.
  • Traceable®
  • Reads °C and °F
  • Triple-Purpose Probe
Minimum and maximum readings may be recalled with this wide-range thermometer
All Type-K probes may be used with liquids, air/gas, and semi-solids. Stainless steel probe with handle has a 15.9 cm (61/4") stem with a 3.3mm (1/8") diameter fitted to an 8.9 cm (31/2") handle. Beaded probe is 1.5mm (1/16") in diameter and features a 1.2m (4') fluoropolymer resin cable. The low-temperature stainless steel probe has a diameter of 4.3mm (3/16") and features a 30.5 cm (12") stem with a 12.7 cm (5") handle. Low-temperature probe is supplied with a 2.4m (8') expandable cable. High-temperature beaded probe is a 3m (10') braided metal wire cable with a smooth tip and a diameter of 5mm (3/16").
LCD display measures 1.3 cm (1/2"). Membrane keypad provides a splashproof design.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate thermometer traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a fast-response Type-K beaded probe (61161-372), and a 9V battery. Additional probes that achieve the high end of the temperature range are available upon request; contact your Avantor sales representative for more information.