Compact Digital Overhead Stirrers, Caframo
Supplier: Caframo
BDC2002AU BDC2002U1 BDC2002 BDC2002U1 BDC2002
10860-014EA 1361.2 USD
10860-014 CA10860-014 77776-540 10860-012 CA58960-174
Compact Digital Overhead Stirrers, Caframo
Stirrers Overhead Stirrers
This Compact Digital Overhead Stirrer maximizes productivity, fits into any area of the laboratory including a fume hood or enclosed workstation, and provides accurate and repeatable results every time.
Suitable for a wide range of mixing applications, this Caframo benchtop overhead stirrers is used in formulations, reaction systems, pharmaceuticals, QA testing, and education classrooms.
With the push of a button, the unique Stirlight™ lights up so that solutions can be seen clearly. The Compact Digital stirrer is powered by a brushless DC (BDC) motor. The belt driven transmission is smooth, quiet and maintenance free. The four button touch control (power on/off, speed increase, speed decrease, light on/off) is easy to use.
Ordering information: Clamp, stand, and impellers are available. Chuck (adjustable up to 10 mm or 3/8 in), chuck key, mounting support rod, external power supply, grounded power cord and manual in four languages (English, French, Spanish and German) are included. Three year warranty.