Accessories for Overhead Stirrers, Caframo
Supplier: Caframo
A120
77776-350CS 607.14 USD
77776-350
Accessories for Overhead Stirrers, Caframo
Stirrers Overhead Stirrers
The Caframo Fastening Clamp attaches the overhead stirrer to the stand
It is composed of a cast zinc aluminum alloy with chemical resistant epoxy coating. The clamp accepts a 1/2" diameter stirrer support rod and stand rods between 0.6 and 1.2" in diameter. Handles are composed of 304 stainless steel.