Safety Stand for Overhead Stirrers, Caframo
Supplier: Caframo
A110
77776-343CS 948.79 USD
77776-343
Safety Stand for Overhead Stirrers, Caframo
Support Stands
The Caframo Safety Stand offers stable support for all Caframo stirrers.
The base is a cast zinc-aluminum alloy coated with chemical resistant epoxy paint and comes with two level adjustment knobs.
Ordering information: Includes the stand base and 28 x 1" dia. 304 stainless steel hollow rod. Additional rod lengths also available.