Mouse Recombinant TH9 CELL POLARIZING PB (from E. coli)
Supplier: VWR International
77753-622
77753-622EA 287.85 USD
77753-622
Mouse Recombinant TH9 CELL POLARIZING PB (from E. coli)
Proteins and Peptides
Cytokines for polarizing mouse T cells into Th9 cells. Th9 cells are a unique subset of T helper cells that produce IL-9. Although the exact function of IL-9 producing T cells is yet to be completely clear it is proposed that these cells could be important in inflammation and helminth infections. Th9 cells can be generated from naive CD4⁺ T cells in culture by culturing them in the presence of IL-4 and TGF-β 1. It also appears that IFNγ and IL-27 can inhibit the generation of Th9 cells.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.