Human Recombinant FGF-8 (from E. coli)
Supplier: VWR International
77753-460 77753-462 77753-464 77753-466 77753-468
77753-460EA 600.95 USD
77753-460 77753-462 77753-464 77753-466 77753-468
Human Recombinant FGF-8 (from E. coli)
Proteins and Peptides
Fibroblast growth factor 8 (FGF-8) is a critical mitogenic factor that is required for normal development of the eye, ear, brain, and limb. FGF-8 functions broadly to promote cell proliferation, differentiation, and migration. Overexpression of FGF-8 increases tumor growth and angiogenesis. Human and mouse FGF-8 proteins show 100% homology.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.