Rat Recombinant FGF-9 (from E. coli )
Supplier:
VWR International
Fibroblast growth factor 9 (FGF-9) is a mitogen and survival factor for nerve and mesenchymal cells. FGF-9 functions as an autocrine and paracrine factor to support the growth and survival of motor neurons and prostate tissue. FGF-9 expression in the gonad is also necessary for sex determination.
High quality
Low endotoxin
Affordable
Caution: For research use only.