Rat Recombinant EGF (from E. coli)
Supplier: VWR International
77752-850 77752-852 77752-854 77752-856 77752-858
77752-850EA 239.88 USD
77752-850 77752-852 77752-854 77752-856 77752-858
Rat Recombinant EGF (from E. coli)
Proteins and Peptides
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR).
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.