Mouse Recombinant IL-11 (from E. coli)
Supplier: VWR International
77752-568 77752-570 77752-572 77752-574
77752-568EA 926.68 USD
77752-568 77752-570 77752-572 77752-574
Mouse Recombinant IL-11 (from E. coli)
Proteins and Peptides
Interleukin 11 (IL-11) is a member of the gp130 family of cytokines. IL-11 functions to promote hematopoietic stem cell proliferation and megakaryocyte differentiation. In non-hematopoietic cell populations, IL-11 stimulates acute-phase proteins, modulates the development of immunoglobulin-producing B cells, and regulates bone turnover. IL-11 binds the IL-11Rα receptor to activate JAK downstream signaling.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.