Mouse Recombinant VEGF-120 (from E. coli)
Supplier: VWR International
77752-430 77752-432 77752-434 77752-436
77752-430EA 696.9 USD
77752-430 77752-432 77752-434 77752-436
Mouse Recombinant VEGF-120 (from E. coli)
Proteins and Peptides
Vascular endothelial growth factor A (VEGF-A) is produced by a wide variety of cell types, including tumor and vascular cells. VEGF-A is a mediator of vascular growth, vascular permeability, and plays a role in stimulating vasodilation via nitric oxide-dependent pathways. Mouse VEGF-A has several alternatively spliced isoforms, with one being VEGF-120. VEGF-120 is an angiogenic factor that is expressed throughout endochondral bone development and is important during skeletogenesis.
  • High quality
  • Low endotoxin
  • Affordable
Caution:
For research use only.