Mouse Recombinant IL-27/P28 (from E. coli)
Supplier: VWR International
77752-414 77752-416 77752-418 77752-412
77752-414EA 262.6 USD
77752-414 77752-416 77752-418 77752-412
Mouse Recombinant IL-27/P28 (from E. coli)
Proteins and Peptides
The p28 subunit of interleukin 27 (IL-27), also known as interleukin 30 (IL-30), is a member of the interleukin 12 (IL-12) family of cytokines. p28 is a secreted polypeptide that associates with the Epstein-Barr virus induced gene 3 (EBI3) to form the IL-27 cytokine heterodimer complex.
IL-27 functions as a proinflammatory cytokine that induces immunomodulatory effects in naïve CD4+ T cells, mast cells, and monocytes. p28 can also form a complex with cytokine-like factor 1 (CLF), that is secreted by dendritic cells, to regulate natural killer (NK) and T cell functions.