Mouse Recombinant TNF-A (from E. coli)
Supplier: VWR International
77752-240 77752-242 77752-244 77752-246 77752-248
77752-240EA 664.08 USD
77752-240 77752-242 77752-244 77752-246 77752-248
Mouse Recombinant TNF-A (from E. coli)
Proteins and Peptides
Tumor necrosis factor alpha (TNF-α) is an inflammatory cytokine secreted by macrophages, monocytes, neutrophils, T cells, and NK-cells following stimulation by bacterial lipopolysaccharide (LPS).  TNF-α signal activation occurs through two receptors, TNFR1 and TNFR2. TNFR1 is expressed on most cell types, unlike TNFR2, which is expressed mainly on immune cells. TNF-α functions to stimulate phagocytosis in macrophages, chemoattract neutrophils, increase insulin resistance, and induce fever.
  • High quality
  • Low endotoxin
  • Affordable
Caution:
For research use only.